missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminodiphenylmethane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC304321000

 View more versions of this product

Catalog No. AC304321000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Aminodiphenylmethane
Authentic
1.5950 to 1.5970 (20°C, 589nm)
Clear colorless to yellow
≥96.0%
97%
(C6H5)2CHNH2
MFCD00008059
15, 1078
91-00-9
Glass bottle
1.0600g/mL
Liquid
100g
C13H13N
183.25
12, 1323
Benzhydrylamine

Chemical Identifiers

97%
(C6H5)2CHNH2
MFCD00008059
≥96.0%
C13H13N
183.25
Aminodiphenylmethane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning