missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminoacetonitrile bisulfate, 98%, Acros Organics™

Manufacturer:  Acros Organics AC153725000

 View more versions of this product

Catalog No. AC153725000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Aminoacetonitrile bisulfate
Glass bottle
123.0°C to 125.0°C
151-63-3
C2H6N2O4S
MFCD00012851
GTGIXCPOLMWQTC-UHFFFAOYSA-N
2-aminoacetonitrile;sulfuric acid
196971
Crystalline Powder and Chunks
Authentic
Beige-Yellow to White
500g
99%
H2NCH2CN·H2SO4
aminoacetonitrile bisulfate, 2-aminoacetonitrile sulfate, aminoacetonitrile hydrogen sulfate, aminoacetonitrile sulfate 1:1, aminoacetonitrile hydrogensulfate, 2-aminoacetonitrile; sulfuric acid, aminoacetonitrile bisulphate, unii-7xa82hgq1c, aminoacetonitrile hydrosulfate, aminomethylcyanide hydrogen sulfate
C(C#N)N.OS(=O)(=O)O
154.14
154.14
98%

Chemical Identifiers

151-63-3
154.14
GTGIXCPOLMWQTC-UHFFFAOYSA-N
196971
C(C#N)N.OS(=O)(=O)O
C2H6N2O4S
MFCD00012851
aminoacetonitrile bisulfate, 2-aminoacetonitrile sulfate, aminoacetonitrile hydrogen sulfate, aminoacetonitrile sulfate 1:1, aminoacetonitrile hydrogensulfate, 2-aminoacetonitrile; sulfuric acid, aminoacetonitrile bisulphate, unii-7xa82hgq1c, aminoacetonitrile hydrosulfate, aminomethylcyanide hydrogen sulfate
2-aminoacetonitrile;sulfuric acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents