Please login to your online account to display your discounted pricing

Aminoacetonitrile bisulfate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Aminoacetonitrile bisulfate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C2H4N2·H2SO4
Linear Formula H2NCH2CN·H2SO4
Formula Weight 154.14
MDL Number MFCD00012851
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC153725000

  SDS
thermo fisher scientific
AC153725000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC153720050

  SDS
thermo fisher scientific
AC153720050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 154.14
Physical Form Crystalline Powder and Chunks
Quantity 500g
Color White to yellow-beige
Linear Formula H2NCH2CN·H2SO4
CAS 151-63-3
Assay Percent Range 99%
Chemical Name or Material Aminoacetonitrile bisulfate
Molecular Formula C2H4N2·H2SO4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00012851
Packaging Glass bottle
Synonym Glycinonitrile bisulfate
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.