Learn More
alpha-Amino-gamma-butyrolactone hydrobromide, 99%, Thermo Scientific Chemicals
$230.08 - $230.08
Chemical Identifiers
CAS | 6305-38-0 |
---|---|
Molecular Formula | C4H7NO2·BrH |
Molecular Weight (g/mol) | 182.02 |
MDL Number | MFCD00012723 |
InChI Key | MKLNTBLOABOJFZ-UHFFFAOYSA-N |
Synonym | alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr |
PubChem CID | 73508 |
IUPAC Name | 3-aminooxolan-2-one;hydrobromide |
SMILES | C1COC(=O)C1N.Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC103290100
|
Thermo Scientific Chemicals
103290100 |
10 g | Glass bottle |
Each for $230.08
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6305-38-0 | |
182.02 | |
MKLNTBLOABOJFZ-UHFFFAOYSA-N | |
73508 | |
C1COC(=O)C1N.Br |
C4H7NO2·BrH | |
MFCD00012723 | |
alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr | |
3-aminooxolan-2-one;hydrobromide |
Specifications
6305-38-0 | |
100.0 | |
White to Beige | |
98.5% min. (Argentometry) | |
C4H7NO2·BrH | |
10 g | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone | |
C1COC(=O)C1N.Br | |
182.02 | |
182.02 | |
Crystalline Powder |
98.5 | |
221.0°C | |
Authentic | |
Glass bottle | |
MFCD00012723 | |
alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr | |
MKLNTBLOABOJFZ-UHFFFAOYSA-N | |
3-aminooxolan-2-one;hydrobromide | |
73508 | |
99% | |
α-Amino-γ-butyrolactone hydrobromide, 99% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 228-614-6
RUO – Research Use Only