missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amikacin disulfate, 50mg/mL in distilled water, sterile-filtered, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J67496AC
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Amikacin Disulfate | |
50mg/mL in distilled water, sterile-filtered | |
39831-55-5 | |
MFCD00167475,MFCD00167475 | |
OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O | |
781.75 | |
25 mL |
Amikacin disulfate | |
Liquid | |
C22H47N5O21S2 | |
FXKSEJFHKVNEFI-GCZBSULCSA-N | |
(2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid) | |
134129615 |
Chemical Identifiers
39831-55-5 | |
781.75 | |
FXKSEJFHKVNEFI-GCZBSULCSA-N | |
(2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid) |
C22H47N5O21S2 | |
MFCD00167475,MFCD00167475 | |
134129615 | |
OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O |
Safety and Handling
EINECSNumber : 254-648-6
RTECSNumber : WK1961200
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C