Please login to your online account to display your discounted pricing

Alfa Aesar™ Amikacin disulfate, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Amikacin disulfate
CAS 39831-55-5
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00167475
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67496AB

  SDS
alfa aesar™
J67496AB
10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67496AC

  SDS
alfa aesar™
J67496AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Amikacin disulfate
Physical Form Liquid
CAS 39831-55-5
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00167475
Storage Note 1 -30° to -10°C
Quantity 10mL
RTECS Number WK1961200
Safety and Handling

-30° to -10°C