missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Amidosulfonic acid, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4451014

 View more versions of this product

Catalog No. AA4451014



Specifications

Specifications

Amidosulfonic acid
Odorless
25g
5329-14-6
H3NO3S
UN2967
amidosulfonic acid
IIACRCGMVDHOTQ-UHFFFAOYSA-N
sulfamic acid
5987
97.09
99.99%
2.12 g/mL
~205°C (decomposition)
1.2
(metals basis)
MFCD00011603
14,8921
Soluble in water.
NS(=O)(=O)O
97.088
CHEBI:9330
Crystalline

Chemical Identifiers

5329-14-6
97.088
IIACRCGMVDHOTQ-UHFFFAOYSA-N
5987
sulfamic acid
H3NO3S
MFCD00011603
amidosulfonic acid
CHEBI:9330
NS(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P234-P264b-P280g-P280i-P302+P352-P305+P351+P338-P332+P313-P362-P501c

H290-H315-H319

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SULFAMIC ACID

EINECSNumber : 226-218-8

RTECSNumber : W05950000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents