missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amidosulfonic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1883636

 View more versions of this product

Catalog No. AAA1883636



Description

Description

Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.

Solubility
Slightly soluble in water, dimethyl fluoride and methanol. Insoluble in hydrocarbons.

Notes
Incompatible with chlorine compounds, nitrates, carbonates, strong oxidizing agents and strong bases.
Specifications

Specifications

1.2
∼205°C (decomposition)
2.12
500 g
H2NSO3H
UN2967
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
IIACRCGMVDHOTQ-UHFFFAOYSA-N
sulfamic acid
5987
97.09
Amidosulfonic acid
5329-14-6
Odorless
H3NO3S
MFCD00011603
148921
Slightly soluble in water,dimethyl fluoride and methanol. Insoluble in hydrocarbons.
NS(=O)(=O)O
97.088
CHEBI:9330
≥98%

Chemical Identifiers

5329-14-6
97.088
IIACRCGMVDHOTQ-UHFFFAOYSA-N
5987
sulfamic acid
H3NO3S
MFCD00011603
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
CHEBI:9330
NS(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P234-P264b-P280g-P280i-P302+P352-P305+P351+P338-P332+P313-P362-P501c

H290-H315-H319

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SULFAMIC ACID

EINECSNumber : 226-218-8

RTECSNumber : WO5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only