Learn More
Amidinothiourea, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 259191000
Specifications
Amidinothiourea | |
0.3% max. | |
White | |
100g | |
2114-02-5 | |
C2H6N4S | |
MFCD00014472 | |
Solubility in water: 7 w/w % in water (20°C) | |
C(=NC(=S)N)(N)N | |
118.16 | |
118.16 | |
99+% |
Authentic | |
Glass bottle | |
160°C to 164°C | |
99+% | |
99% min. (Iodimetry) | |
NH2C(=NH)NHC(=S)NH2 | |
2-imino-4-thiobiuret, amidinothiourea, diaminomethylidene thiourea, guanylthiourea, guthimin, guthimine, gutimine, n-amidinothiourea, thiodicyanodiamidine, thiourea, aminoiminomethyl | |
OKGXJRGLYVRVNE-UHFFFAOYSA-N | |
diaminomethylidenethiourea | |
2724563 | |
Crystalline Powder or Crystals |
Chemical Identifiers
2114-02-5 | |
118.16 | |
OKGXJRGLYVRVNE-UHFFFAOYSA-N | |
2724563 | |
C(=NC(=S)N)(N)N |
C2H6N4S | |
MFCD00014472 | |
2-imino-4-thiobiuret, amidinothiourea, diaminomethylidene thiourea, guanylthiourea, guthimin, guthimine, gutimine, n-amidinothiourea, thiodicyanodiamidine, thiourea, aminoiminomethyl | |
diaminomethylidenethiourea |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 218-308-0
RTECSNumber : YR8250000