missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amidinothiourea, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 259190100

 View more versions of this product

Catalog No. AC259190100



Description

Description

Specifications

Specifications

Amidinothiourea
0.3% max.
White
10g
2114-02-5
C2H6N4S
MFCD00014472
Solubility in water: 7 w/w % in water (20°C)
C(=NC(=S)N)(N)N
118.16
118.16
99+%
Authentic
Glass bottle
160°C to 164°C
99+%
99% min. (Iodimetry)
NH2C(=NH)NHC(=S)NH2
2-imino-4-thiobiuret, amidinothiourea, diaminomethylidene thiourea, guanylthiourea, guthimin, guthimine, gutimine, n-amidinothiourea, thiodicyanodiamidine, thiourea, aminoiminomethyl
OKGXJRGLYVRVNE-UHFFFAOYSA-N
diaminomethylidenethiourea
2724563
Crystalline Powder or Crystals

Chemical Identifiers

2114-02-5
118.16
OKGXJRGLYVRVNE-UHFFFAOYSA-N
2724563
C(=NC(=S)N)(N)N
C2H6N4S
MFCD00014472
2-imino-4-thiobiuret, amidinothiourea, diaminomethylidene thiourea, guanylthiourea, guthimin, guthimine, gutimine, n-amidinothiourea, thiodicyanodiamidine, thiourea, aminoiminomethyl
diaminomethylidenethiourea
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-308-0

RTECSNumber : YR8250000

SDS Documents