missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AMI-193, Tocris

D2-like receptor antagonist

$181.19 - $762.58

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C22H26FN3O2
Formula Weight 383.46
Synonym 8-[3-(4-Fluorophenoxy)propyl]-1-phenyl-1, 3, 8-triazaspiro[4.5]-decanone
Chemical Name or Material AMI-193
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
052410 R & D Systems
RDS052410
10mg Each for $181.19
052450 R & D Systems
RDS052450
50mg Each for $762.58
Specifications Description & Specifications

Chemical Identifiers

>99%
383.46
AMI-193
C22H26FN3O2
8-[3-(4-Fluorophenoxy)propyl]-1-phenyl-1, 3, 8-triazaspiro[4.5]-decanone

Specifications

10mg
>99%
383.46
AMI-193
510-74-7
C22H26FN3O2
8-[3-(4-Fluorophenoxy)propyl]-1-phenyl-1, 3, 8-triazaspiro[4.5]-decanone

Safety and Handling

Recommended Storage : Store at Room Temperature