Please login to your online account to display your discounted pricing

AMI-193, Tocris


D2-like receptor antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS052450

 View more versions of this product

Catalog No. 052450

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material AMI-193
Quantity 50mg
CAS 510-74-7
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C22H26FN3O2
Formula Weight 383.46
Synonym 8-[3-(4-Fluorophenoxy)propyl]-1-phenyl-1, 3, 8-triazaspiro[4.5]-decanone

Specifications

Chemical Name or Material AMI-193
Assay Percent Range >99%
Molecular Formula C22H26FN3O2
Formula Weight 383.46
Synonym 8-[3-(4-Fluorophenoxy)propyl]-1-phenyl-1, 3, 8-triazaspiro[4.5]-decanone
Accessories

Store at Room Temperature