Please login to your online account to display your discounted pricing

AMG 9810, Tocris

Potent and selective, competitive antagonist of TRPV1

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material AMG 9810
Assay Percent Range >98%
Molecular Formula C21H23NO3
Formula Weight 337.42
Synonym (2E)-N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-[4-(1,1-dimethylethyl)phenyl]-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

231610

 
r & d systems
RDS231610
10mg Each for $221.01

231650

 
r & d systems
RDS231650
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 337.42
Quantity 10mg
CAS 545395-94-6
Molecular Formula C21H23NO3
Chemical Name or Material AMG 9810
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C
Synonym (2E)-N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-[4-(1,1-dimethylethyl)phenyl]-2-propenamide
Safety and Handling

Desiccate at +4°C