missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-AMG 487, Tocris™

CXCR3 antagonist; inhibits cell migration and metastasis

$303.99 - $1,257.94

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C32H28F3N5O4
Formula Weight 603.59
Synonym N-1-[(3-4(-Ethoxyphenyl)-3, 4-dihydro-4-oxopyrido[2, 3-d]pyrimidin-2-yl]ethyl]-N-(3-pyridinylmethyl)-4-(trifluoromethoxy)benzeneacetamide
Chemical Name or Material (±)-AMG 487
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
448710 R & D Systems
448710
10mg Each for $303.99
448750 R & D Systems
448750
50mg Each for $1,257.94
Specifications Description & Specifications

Chemical Identifiers

>98%
603.59
(±)-AMG 487
C32H28F3N5O4
N-1-[(3-4(-Ethoxyphenyl)-3, 4-dihydro-4-oxopyrido[2, 3-d]pyrimidin-2-yl]ethyl]-N-(3-pyridinylmethyl)-4-(trifluoromethoxy)benzeneacetamide

Specifications

10mg
C32H28F3N5O4
N-1-[(3-4(-Ethoxyphenyl)-3, 4-dihydro-4-oxopyrido[2, 3-d]pyrimidin-2-yl]ethyl]-N-(3-pyridinylmethyl)-4-(trifluoromethoxy)benzeneacetamide
>98%
603.59
(±)-AMG 487

Safety and Handling

Recommended Storage : Store at -20°C