missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Amersham™ ECL™ Blocking Agent PROMO

Use for blocking nitrocellulose and PVDF membranes. Amersham™ ECL™ Blocking Agent blocks membranes to be used with ECL and ECL Plus™ Western blotting detection reagents.

Manufacturer:  Cytiva 29822203

Catalog No. 45001197LR2



Description

Description

  • Contains 40 g of membrane blocking agent
  • Sufficient for at least 20 miniblots
Specifications

Specifications

40 g
40 g ECL™ blocking agent, sufficient for at least 20 miniblots
Blocking Nitrocellulose and PVDF membranes to be used with ECL™ and ECL™ PlusTM western blotting detection reagents
SDS
Documents

Documents

Product Certifications
Promotions

Promotions