missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
American Fiber and Finishing Purewipe™ Washable Cheesecloth Wipers
For cleaning, polishing, straining, and filtering
Manufacturer: American Fiber and Finishing 588036
Description
Washable. Grade 40: fine weave, 24 × 20 threads/sq. in.
- 100% cotton
- Biodegradable and incinerable
- Silicone-free
- No ozone-depleting agents
Order Info
Cut off what you need. In handifold bolt of 70 yd. (64m).
Specifications
Cleaning, polishing, straining, and filtering | |
Woven Cotton | |
Purewipe™ Washable cheesecloth wipes |