Please login to your online account to display your discounted pricing

American Educational Products Test Tube Tongs

Brass clamp won't rust or corrode.

$7.50 - $8.35

Specifications

Material Brass
Type Test Tube Tong
For Use With (Application) Provides long dependable service for carrying 0.25 x 1.25 in. dia. tubes
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity    

S49062

 
american educational products
H8595
11.5 in. 29.2cm Each for $7.50
Pack of 10 Each for $74.80

S90723

 
american educational products
7-G73
5 in. 12.7cm Each for $8.35
Description & Specifications

Specifications

Material Brass
Type Test Tube Tong
For Use With (Application) Provides long dependable service for carrying 0.25 x 1.25 in. dia. tubes

  • Fit 0.25 × 1.25 in. (0.6 to 3.1cm) dia. tubes
  • Offer long, dependable service
  • Choose tongs with or without finger grips
  • Length: with grips, 5 in. (12.7cm); without grips, 5.5 in. (13.75cm)