American Educational Products Students' Voltaic Cell with Electrodes
For studying electrochemical reactions.
$5.35 - $5.35
Specifications
Category Physics
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Product Type Price Quantity  
S04328A American Educational Products
7-1826-PC
Designed to hold both round and flat electrodes Porous Cup Each for $5.35
Specifications
Description
Designed to hold both round and flat electrodes.
  • With eight electrodes
  • Top rim is made of molded plastic
  • Nickel-plated clamps
  • Designed to hold both round and flat electrodes
Order Info
Diameter × H: 4 × 5 in. (95 × 120mm); weight: 6 lb. (2.7kg).
Description & Specifications
Specifications
Physics