American Educational Products Steel Motion Balls
Drilled steel alloy.
Manufacturer:  American Educational Products 20041"
 View more versions of this product
Catalog No. S40975
Description
Description
  • Used in collision and pendulum activities, as well as other force, motion, and mass demonstrations
  • Select from balls with 3/4 or 1 in. holes
Specifications
Specifications
Collision and pendulum activities, as well as other force, motion, and mass demonstrations
Steel
0.22 lb.
1 in. (25.4mm) hole
Motion Balls
Documents