Please login to your online account to display your discounted pricing
American Educational Products Steel Motion Balls
Drilled steel alloy.
$5.35 - $10.35
Specifications
Product Type Motion Balls
For Use With (Application) Collision and pendulum activities, as well as other force, motion, and mass demonstrations
Material Steel
View More Specs
Products
Catalog Number Mfr. No. Includes Weight (English) Price Quantity    
S40974
american educational products
20043/4"
3/4 in. (19mm) hole 0.16 lb. Each for $5.35
Pack of 10 Each for $53.20
S40975
american educational products
20041"
1 in. (25.4mm) hole 0.22 lb. Each for $10.35
Pack of 10 Each for $103.55
Description & Specifications
Specifications
Product Type Motion Balls
For Use With (Application) Collision and pendulum activities, as well as other force, motion, and mass demonstrations
Material Steel
  • Used in collision and pendulum activities, as well as other force, motion, and mass demonstrations
  • Select from balls with 3/4 or 1 in. holes