Please login to your online account to display your discounted pricing
American Educational Products Molecular Model Kits for Organic Chemistry
Hands-on modeling of organic chemistry compounds.
Manufacturer: american educational products  1864
 View all options for this product
Catalog No. S67964
Description & Specifications
Includes Colored balls in two sizes (49), white Hydrogen atoms with bonds (25), bonding lugs in the three sizes mentioned above (30), clear Plastic storage case and an instruction manual
Type American educational products molecular model kit for organic chemistry
  • Unbreakable plastic balls are precision drilled to ensure proper bond angles
  • Tapered plastic bonding rods simplify the construction process
  • Varying lengths of bonding rods represent the relative bonding lengths that occur between molecules
  • 1 in. rods (2.54cm) represent the Carbon-Carbon alkane bond; 0.75 in. rods (1.9cm) represent double and triple bonding
  • 2 in. spare rods (5.1cm) can be cut to any desired length