American Educational Products Molecular Model Kits for Organic Chemistry
Hands-on modeling of organic chemistry compounds.
Manufacturer:  American Educational Products 1863*
 View more versions of this product
Catalog No. S67963
Description
Description
  • Unbreakable plastic balls are precision drilled to ensure proper bond angles
  • Tapered plastic bonding rods simplify the construction process
  • Varying lengths of bonding rods represent the relative bonding lengths that occur between molecules
  • 1 in. rods (2.54cm) represent the Carbon-Carbon alkane bond; 0.75 in. rods (1.9cm) represent double and triple bonding
  • 2 in. spare rods (5.1cm) can be cut to any desired length
Specifications
Specifications
1 in. and 0.75 in. colored balls (245), white Hydrogen atoms with bonds (125), 1 in. bonding rods (90), 0.75 in. bonding rods (60), 2 in. bonding rods (10), cardboard container with separate, compartments for each color and an instruction manual
American educational products molecular model kit for organic chemistry
Documents