Please login to your online account to display your discounted pricing
American Educational Products Molecular Model Kits for Organic Chemistry
Hands-on modeling of organic chemistry compounds.
$438.75 - $438.75
Specifications
Type American educational products molecular model kit for organic chemistry
View More Specs
Includes: 
Instructor Set: 1 in. and 0.75 in. (2.54, 1.9cm) colored balls (245), white Hydrogen atoms with bonds (125), 1 in. (2.54cm) bonding rods (90), 0.75 in. (1.9cm) bonding rods (60), 2 in. (5.1cm) bonding rods (10), cardboard container with separate compartments for each color, and an instruction manual
Student Set: Colored balls in two sizes (49), white Hydrogen atoms with bonds (25), bonding lugs in the three sizes mentioned above (30), clear plastic storage case and an instruction manual
Products
Catalog Number Mfr. No. Includes Price Quantity    
S67963
american educational products
1863*
1 in. and 0.75 in. colored balls (245), white Hydrogen atoms with bonds (125), 1 in. bonding rods (90), 0.75 in. bonding rods (60), 2 in. bonding rods (10), cardboard container with separate, compartments for each color and an instruction manual Each for $438.75
Case of 10 Each for $4,387.50
Description & Specifications
Specifications
Type American educational products molecular model kit for organic chemistry
  • Unbreakable plastic balls are precision drilled to ensure proper bond angles
  • Tapered plastic bonding rods simplify the construction process
  • Varying lengths of bonding rods represent the relative bonding lengths that occur between molecules
  • 1 in. rods (2.54cm) represent the Carbon-Carbon alkane bond; 0.75 in. rods (1.9cm) represent double and triple bonding
  • 2 in. spare rods (5.1cm) can be cut to any desired length