Please login to your online account to display your discounted pricing
American Educational Products Miniature Lamps, Screw Base
Use to demonstrate many simple circuits.
Manufacturer: american educational products  605-PC26551
 View more versions of this product
Catalog No. S43908
  • / Pack of 10 Each
  • / Case of 50 Pack
Description & Specifications
Specifications
Voltage 6.3V
Lighting Type Miniature Lamp
Includes Screw base
  • Serve as well as large lamps to demonstrate many simple circuits
  • No hazard with low-voltage source; low current drain won't overload batteries
  • Sold in packs of 10