Please login to your online account to display your discounted pricing
American Educational Products Knife Switches
These knife switches are great for classroom and laboratory experiments.
Manufacturer: american educational products  9918001
 View more versions of this product
Catalog No. S44402
  • / Each
  • / Pack of 10 Each
Description & Specifications
Specifications
Includes Double pole, single throw
Product Type Knife Switch
For Use With (Application) Classroom and laboratory experiments, each has a mounted screw binding post for a solid electrical connection