American Educational Products Jewel PlantMobile
Durable construction, designed for mobility.
Manufacturer:  American Educational Products 52032
 View more versions of this product
Catalog No. S17618MF
Specifications Description & Specifications
Specifications
Jewel 56 in. PlantMobile
56 in.
52 x 19 x 56 in. (132 x 48.26 x 142.24cm)
132cm
52 in.
142.24cm
Two adjustable UL-approved lighthoods, three open shelves with three 18 x 47 in. Plastic growing trays, assorted pots, 3 in. locking wheels, teacher's manual
19 in.
UL
48.26cm