Please login to your online account to display your discounted pricing
American Educational Products Iron Triangles
Budget-priced triangles for heating crucibles.
Manufacturer: american educational products  8026EMD
 View more versions of this product
Catalog No. S48062
  • / Pack of 12 Each
Description & Specifications
Specifications
Product Type Triangle
For Use With (Equipment) Heating crucibles
Includes Clay tube
Length (English) 2.5 in.
Length (Metric) 6.4cm
Material Iron Wire