Please login to your online account to display your discounted pricing
American Educational Products Investigating Measurements and Density Unit
Two-in-one kit covers both measurement and density.
$34.10 - $202.50
Specifications
Grade 6 to 12
View More Specs
Includes: 
  • Ruler
  • Measuring cylinders
  • Mass pieces
  • Wooden and Plexiglas™ blocks
  • Workbooks: Investigating Measurement and Investigating Density
Requires:  School balance
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S19405
american educational products
4000701
Investigating measurements workbook Pack of 10 Each for $34.10
S19423
american educational products
R100101
Investigating measurement and density kit Each for $202.50
S19418
american educational products
4000702
Investigating density workbook Pack of 10 Each for $34.10
Description & Specifications
Specifications
Grade 6 to 12
  • Measure length, temperature, mass and gas volume
  • Calculate the volume of regular and irregular solids
  • Determine the density of water and several irregular solids
  • Covers 18 scientific investigations
  • Additional workbooks available separately