Please login to your online account to display your discounted pricing
American Educational Products High-Form Crucibles
Highly resistant to alkalis and high temperatures.
$45.85 - $45.85
Specifications
Product Type Crucible
Finish Glazed
Material Porcelain
With Lid cover
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Height (English) Price Quantity    
S326031
american educational products
C18
0.34 oz. 1.18 in. 1.1 in. Pack of 10 Each for $45.85
Description & Specifications
Specifications
Product Type Crucible
Finish Glazed
Material Porcelain
With Lid cover
  • Glazed inside and out except for outside bottom surface
  • Packaged to prevent breakage and keep the product clean
  • Cover included