Please login to your online account to display your discounted pricing
American Educational Products Graduated Polypropylene Cylinders
Raised outer graduations to facilitate easy reading.
Manufacturer: american educational products  200-500
 View all options for this product
Catalog No. S95176
Description & Specifications
Capacity (Metric) 500mL
Material Polypropylene
Autoclavable Yes
Base Type Broad
  • Constructed of durable polypropylene
  • Broad-based for stability
  • Autoclavable