Please login to your online account to display your discounted pricing

American Educational Products Graduated Polypropylene Beakers


Easy to read.

Manufacturer: american educational products  7-S95154

 View more versions of this product

Catalog No. S95154

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Material PP (Polypropylene)
Capacity 3.38 oz., 100mL
Autoclavable Yes
Color Translucent
For Use With (Application) For pouring large volumes, chemical resistant
Graduated Yes
Includes Spout
Scale Raised outer graduated
Capacity (Metric) 100mL
Product Type Beaker

Tough, translucent polypropylene beakers incorporate raised outer graduations to facilitate easy reading.

  • Pouring spout
  • Chemically resistant
  • Autoclavable