Please login to your online account to display your discounted pricing

American Educational Products Graduated Polypropylene Beakers

Easy to read.

$21.20 - $135.15

Specifications

Autoclavable Yes
Color Translucent
Material PP (Polypropylene)
For Use With (Application) For pouring large volumes, chemical resistant
Graduated Yes
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    

S95153

 
american educational products
7-100-050
50mL Pack of 12 Each for $21.20

S95158

 
american educational products
100-2000
2000mL Pack of 12 Each for $135.15
Description & Specifications

Specifications

Autoclavable Yes
Color Translucent
Material PP (Polypropylene)
For Use With (Application) For pouring large volumes, chemical resistant
Graduated Yes
Includes Spout
Scale Raised outer graduated

Tough, translucent polypropylene beakers incorporate raised outer graduations to facilitate easy reading.

  • Pouring spout
  • Chemically resistant
  • Autoclavable