Please login to your online account to display your discounted pricing

American Educational Products Graduated Polypropylene Beakers


Easy to read.

Manufacturer: american educational products  100-SET

 View more versions of this product

Catalog No. S95159

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Capacity (Metric) One each: 50, 100, 250, 500, 1000mL
Product Type Beaker
Material PP (Polypropylene)
Includes Spout
Graduated Yes
Autoclavable Yes
Color Translucent
For Use With (Application) For pouring large volumes, chemical resistant
Scale Raised outer graduated
Capacity 50, 100, 250, 500, and 1000mL