Please login to your online account to display your discounted pricing
American Educational Products Glass Plates
For covering jars and beakers.
$21.30 - $21.30
Specifications
For Use With (Application) Used for observing potassium flames
Material Glass
Sterile Yes
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity    
S50345
american educational products
13730A
2.99 in. 7.6cm 2.99 in. 7.6cm Pack of 12 Each for $21.30
Description & Specifications
Specifications
For Use With (Application) Used for observing potassium flames
Material Glass
Sterile Yes
  • Square with unground edges
  • More durable than plastic
  • Easily sterilized
  • Available in brilliant cobalt blue glass for observing potassium flames