Please login to your online account to display your discounted pricing

American Educational Products Flint Glass Wide-Mouth Jars


Lighter and more economical.

Manufacturer: american educational products  2097

 View more versions of this product

Catalog No. S31834

  • / Pack of 12 Each
  • / Case of 6 Pack
 

Description & Specifications

Specifications

Mouth Wide
Capacity (Metric) 237mL
Material Flint Glass
Capacity (English) 8 oz.
Capacity 8 oz., 237mL