Please login to your online account to display your discounted pricing
American Educational Products Flint Glass Wide-Mouth Jars
Lighter and more economical.
$49.30 - $76.95
Specifications
Material Flint Glass
Mouth Wide
View More Specs
Products
Catalog Number Mfr. No. Capacity Price Quantity    
S31834
american educational products
2097
8 oz., 237mL Pack of 12 Each for $49.30
Case of 6 Pack for $295.65
S31835
american educational products
CTT6732484
16 oz., 473mL Pack of 12 Each for $76.95
Case of 4 Pack for $307.80
Description & Specifications
Specifications
Material Flint Glass
Mouth Wide
Wide-mouth, machine-made, nonporous flint glass bottles.
  • Lighter than handmade bottles and less expensive
  • Suitable for gas generation and collection
  • Easily cleaned and autoclavable