Please login to your online account to display your discounted pricing
American Educational Products Flexible Banana Plug Lead Wires
Complete wiring projects quickly and safely.
$6.65 - $6.65
Specifications
Product Type Banana Plug Lead Wire
Length (English) 19.7 in.
Length (Metric) 50cm
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
S44323A
american educational products
ECF310070B
Black Each for $6.65
Description & Specifications
Specifications
Product Type Banana Plug Lead Wire
Length (English) 19.7 in.
Length (Metric) 50cm
  • Copper conductors ensure good continuity
  • PVC-insulated for good short-circuit protection
  • Cluster-type leads with stackable heads have been voltage-tested to 3kV