American Educational Products Flask-Type Student Electroscope
For individual electrostatic investigations.
$28.05 - $28.05
Specifications
For Use With (Application) Individual electrostatic investigations
View More Specs
Includes: Instructions
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S43262 American Educational Products
EMI3420
Flask type electroscope Each for $28.05
Specifications
Description
  • Flask electroscope consists of aluminum leaves mounted on metal rod
  • Rubber stopper holds the rod in place inside a 250mL glass flask
Order Info
Dimensions: 3.5 × 3.5 × 6.5 in. (8.8 × 8.8 × 16.3cm); weight: 0.5 lb. (227g).
Description & Specifications
Specifications
Individual electrostatic investigations