Please login to your online account to display your discounted pricing

American Educational Products Electrical Bench Meters


Designed in a wide array of options to fit your needs.

Manufacturer: american educational products  P57031

 View more versions of this product

Catalog No. S77840

  • / Each
 

Description & Specifications

Specifications

Color Red
Product Type Bench DC ammeter
Includes Insulated thumbscrew
Quantity Each
Style Closed Back
For Use With (Application) Ideal for student use
Scale Single
Amperage 0 to 1A

  • Low-cost,  high-quality  meters  for  numerous  uses
  • Closed back