Please login to your online account to display your discounted pricing

American Educational Products Electrical Bench Meters

Designed in a wide array of options to fit your needs.

Manufacturer: american educational products  P57051

 View all options for this product

Catalog No. S77820


Description & Specifications


Amperage 0 to 100μA
Color Red
Quantity Each
For Use With (Application) Ideal for student use
Product Type Bench DC microammeter
Includes Insulated thumbscrew
Scale Single
Style Closed Back

  • Low-cost,  high-quality  meters  for  numerous  uses
  • Closed back