Please login to your online account to display your discounted pricing

American Educational Products Electrical Bench Meters

Designed in a wide array of options to fit your needs.

Manufacturer: american educational products  P57016

 View all options for this product

Catalog No. S77812


Description & Specifications


Voltage 0 to 10V
Color Blue
Quantity Each
For Use With (Application) Ideal for student use
Product Type Bench voltmeter
Includes Sturdy binding posts for wire attachment
Scale Single

  • Low-cost,  high-quality  meters  for  numerous  uses
  • Closed back