Please login to your online account to display your discounted pricing
American Educational Products Electrical Bench Meters
Designed in a wide array of options to fit your needs.
Manufacturer: american educational products  P27037
 View more versions of this product
Catalog No. S77843
  • / Each
  • / Case of 40 Each
Description & Specifications
Specifications
For Use With (Application) Ideal for student use
Quantity Each
Scale Single
Style Closed Back
Amperage 0 to 10A
Includes Insulated thumbscrew
Product Type Bench DC ammeter
  • Low-cost,  high-quality  meters  for  numerous  uses
  • Closed back