Please login to your online account to display your discounted pricing

American Educational Products Durable Gauze

With ceramic fiber centers for thermal protection.

Manufacturer: american educational products  7-4000-3

 View all options for this product

Catalog No. S95379


Description & Specifications


Quantity 10 Pack
Length (English) 6 in.
Length (Metric) 15cm
Width (English) 6 in.
Material Iron
Product Type Wire Gauze
Width (Metric) 15cm
For Use With (Application) For thermal protection
Dimensions (L x W) 6 x 6 in. (15 x 15cm)

  • Made of woven iron wire
  • Turned edges prevent fraying