Please login to your online account to display your discounted pricing
American Educational Products Durable Gauze
With ceramic fiber centers for thermal protection.
$24.80 - $35.10
Specifications
Material Iron
Product Type Wire Gauze
For Use With (Application) For thermal protection
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity    
S95379
american educational products
7-4000-3
6 x 6 in. (15 x 15cm) Pack of 10 Each for $24.80
S95378
american educational products
7-S95378
5 x 5 in. (13 x 13cm) Pack of 20 Each for $35.10
Description & Specifications
Specifications
Material Iron
Product Type Wire Gauze
For Use With (Application) For thermal protection
  • Made of woven iron wire
  • Turned edges prevent fraying