Please login to your online account to display your discounted pricing

American Educational Products Dropping Bottles


Store reagent solutions in these sturdy dropping bottles.

Manufacturer: american educational products  405-2-DZ

 View more versions of this product

Catalog No. S31681

  • / Pack of 12 Each
  • / Case of 12 Pack
 

Description & Specifications

Specifications

Color Amber
Capacity (Metric) 15mL
Capacity (English) 0.8 oz.
Closure Screw
Includes Dropper assembly
Material Plastic
Shape Round
Capacity 0.8 oz., 15mL