Please login to your online account to display your discounted pricing

American Educational Products Dropping Bottles


Store reagent solutions in these sturdy dropping bottles.

Manufacturer: american educational products  S31678

 View more versions of this product

Catalog No. S31678

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Capacity (Metric) 250mL
Capacity (English) 8.45 oz.
Closure Screw
Color Clear
Includes Dropper assembly
Material Glass
Shape Round
Capacity 8.45 oz., 250mL