Please login to your online account to display your discounted pricing

American Educational Products Dropping Bottles


Store reagent solutions in these sturdy dropping bottles.

Manufacturer: american educational products  4091ADZ

 View more versions of this product

Catalog No. S31672

  • / Pack of 12 Each
  • / Case of 12 Pack
 

Description & Specifications

Specifications

Closure Screw
Capacity (Metric) 30mL
Capacity (English) 1 oz.
Color Clear
Includes Dropper assembly
Material Plastic
Shape Round
Capacity 1 oz., 30mL