Please login to your online account to display your discounted pricing
American Educational Products Dissection Trays
Ideal for general student use.
Manufacturer: american educational products  7-350
 View more versions of this product
Catalog No. S65105
  • / Each
Description & Specifications
Specifications
Material Aluminum
Type Standard Tray
Durable, one-piece aluminum dissection trays won't rust, crack or leak.
  • Standard-size tray measures 11.5 × 7.5 in. (28.8 × 18.9cm)
  • Deluxe tray measures 13.13 × 9.38 × 2.25 in. (33 × 23.5 × 5.6cm)
  • Both come lined with black wax that can be reused or resurfaced for continual use