Please login to your online account to display your discounted pricing

American Educational Products Connector Cords and Alligator Clips


Safe and easy for students to use.

Manufacturer: american educational products  7-199-19-B

 View more versions of this product

 Alert

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S95303

  • / Each
 

Description & Specifications

Specifications

Product Type Alligator Clip
For Use With (Application) For students to use
Length (English) 12 in.
Color Black
Length (Metric) 30.5cm
Material Plastic