Please login to your online account to display your discounted pricing
American Educational Products Connector Cords and Alligator Clips
Safe and easy for students to use.
$3.30 - $29.10
Specifications
For Use With (Application) For students to use
Material Plastic
View More Specs
 Alert
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Products
Catalog Number Mfr. No. Color Length (English) Length (Metric) Price Quantity    
S95302
american educational products
7-199-19-R
Red 12 in. 30.5cm Each for $3.40
S95304
american educational products
7-199-21-R
Red 24 in. 61cm Each for $4.25
S95303
american educational products
7-199-19-B
Black 12 in. 30.5cm Each for $3.30
S95305
american educational products
7-199-21-B
Black 24 in. 61cm Each for $4.25
S95302PK
american educational products
7-S95302
Red 12 in. 30.5cm Pack of 12 Each for $28.40
S95303PK
american educational products
7-S95303
Black 12 in. 30.5cm Pack of 12 Each for $29.10
Description & Specifications
Specifications
For Use With (Application) For students to use
Material Plastic
  • Teeth on alligator clips maintain solid contact
  • Banana plugs provide good continuity
  • Spade wires simplify wiring, and the flexible cords are plastic-coated to ensure added safety in the classroom